Learn More
Biolegend Purified anti-human Granulysin
Supplier:  Biolegend 348007
Encompass_Preferred
Purified anti-human Granulysin [DH2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IHC-F; Size: 50 ug
Catalog No. 50-207-6842