Learn More
Biolegend Purified anti-human Folate Receptors α and β (FR-αβ)
Supplier:  Biolegend 391802
Encompass_Preferred
Purified anti-human Folate Receptors α and β (FR-αβ) [No.5/FOLR]; Isotype: Rat IgG2a, κ; Reactivity: Human, Monkey; Apps: FC; Size: 100 ug
Catalog No. 50-207-8874