Learn More
Biolegend Purified anti-human β2-microglobulin
Supplier:  Biolegend 316302
Encompass_Preferred
Purified anti-human β2-microglobulin [2M2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC, WB, ELISA; Size: 100 ug
Catalog No. 50-166-855