Learn More
Biolegend Purified anti-human CD275 (B7-H2, ICOSL)
Supplier:  Biolegend 309402
Encompass_Preferred
Purified anti-human CD275 (B7-H2, ICOSL) [2D3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC, IP, ELISA; Size: 100 ug
Catalog No. 50-166-508