Learn More
Biolegend Purified anti-human Folate Receptor β (FR-β)
Supplier:  Biolegend 391702
Encompass_Preferred
Purified anti-human Folate Receptor β (FR-β) [94b/FOLR2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-8868