Learn More
Biolegend Purified anti-human CD10 (Maxpar Ready)
Supplier:  Biolegend 312223
Encompass_Preferred
Purified anti-human CD10 (Maxpar Ready) [HI10a]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin monkey, Chimpanzee, Cynomolgus, Rhesus; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-207-3557