Learn More
Biolegend Purified anti-human CD106
Supplier:  Biolegend 305802
Encompass_Preferred
Purified anti-human CD106 [STA]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-P, ICC, IP, ELISA; Size: 100 ug
Catalog No. 50-166-298