Learn More
Biolegend Purified anti-human CD112 (Nectin-2)
Supplier:  Biolegend 337402
Encompass_Preferred
Purified anti-human CD112 (Nectin-2) [TX31]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-167-911