Learn More
Biolegend Purified anti-human CD116
Supplier:  Biolegend 305902
Encompass_Preferred
Purified anti-human CD116 [4H1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC, IP, WB, IHC-P, IHC-F; Size: 100 ug
Catalog No. 50-166-307