Learn More
Biolegend Purified anti-human CD11b
Supplier:  Biolegend 301302
Encompass_Preferred
Purified anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC, CyTOF, ICC, IHC; Size: 100 ug
Catalog No. 50-165-686