Learn More
Biolegend Purified anti-human CD127 (IL-7Rα) Recombinant
Supplier:  Biolegend 375702
Encompass_Preferred
Purified anti-human CD127 (IL-7Rα) Recombinant [QA18A44]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-8844