Learn More
Biolegend Purified anti-human CD13
Supplier:  Biolegend 301702
Encompass_Preferred
Purified anti-human CD13 [WM15]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cotton-topped Tamarin; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-165-735