Learn More
Biolegend Purified anti-human CD137L (4-1BB Ligand)
Supplier:  Biolegend 311502
Encompass_Preferred
Purified anti-human CD137L (4-1BB Ligand) [5F4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-166-649