Learn More
Biolegend Purified anti-human CD14
Supplier:  Biolegend 325602
Encompass_Preferred
Purified anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-167-288