Learn More
Biolegend Purified anti-human CD14
Supplier:  Biolegend 301802
Encompass_Preferred
Purified anti-human CD14 [M5E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Capuchin Monkey, Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Dog (Canine), Rhesus, Pigtailed Macaque, Squirrel Monkey; Apps: FC, CyTOF, Block, IHC-F; Size: 100 ug
Catalog No. 50-165-746