Learn More
Biolegend Purified anti-human CD147
Supplier:  Biolegend 306202
Encompass_Preferred
Purified anti-human CD147 [HIM6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Capuchin Monkey; Apps: FC, IHC-P; Size: 100 ug
Catalog No. 50-166-325