Learn More
Biolegend Purified anti-human CD15 (SSEA-1)
Supplier:  Biolegend 301902
Encompass_Preferred
Purified anti-human CD15 (SSEA-1) [HI98]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, IHC-P, IHC-F; Size: 100 ug
Catalog No. 50-165-771