Learn More
Biolegend Purified anti-human CD155 (PVR)
Supplier:  Biolegend 337602
Encompass_Preferred
Purified anti-human CD155 (PVR) [SKII.4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, WB, IP; Size: 100 ug
Catalog No. 50-167-919