Learn More
Biolegend Purified anti-human CD156c (ADAM10)
Supplier:  Biolegend 352702
Encompass_Preferred
Purified anti-human CD156c (ADAM10) [SHM14]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-168-728