Learn More
Biolegend Purified anti-human CD158 (KIR2DL1/S1/S3/S5)
Supplier:  Biolegend 339502
Encompass_Preferred
Purified anti-human CD158 (KIR2DL1/S1/S3/S5) [HP-MA4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus; Apps: FC, IP; Size: 100 ug
Catalog No. 50-168-001