Learn More
Biolegend Purified anti-human CD170 (Siglec-5)
Supplier:  Biolegend 352002
Encompass_Preferred
Purified anti-human CD170 (Siglec-5) [1A5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ELISA; Size: 100 ug
Catalog No. 50-168-691