Learn More
Biolegend Purified anti-human CD180 (RP105)
Supplier:  Biolegend 312902
Encompass_Preferred
Purified anti-human CD180 (RP105) [MHR73-11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Cynomolgus, African Green, Rhesus; Apps: FC,ICFC, IHC-F, IP; Size: 100 ug
Catalog No. 50-166-706