Learn More
Biolegend Purified anti-human CD184 (CXCR4)
Supplier:  Biolegend 306502
Encompass_Preferred
Purified anti-human CD184 (CXCR4) [12G5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Sooty Mangabey; Apps: FC, ICC, CyTOF, IHC; Size: 100 ug
Catalog No. 50-166-349