Learn More
Biolegend Purified anti-human CD1a
Supplier:  Biolegend 300102
Encompass_Preferred
Purified anti-human CD1a [HI149]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-165-495