Learn More
Biolegend Purified anti-human CD1c
Supplier:  Biolegend 331501
Encompass_Preferred
Purified anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-167-650