Learn More
Biolegend Purified anti-human CD206 (MMR)
Supplier:  Biolegend 321102
Encompass_Preferred
Purified anti-human CD206 (MMR) [15-2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, WB, ICC, IHC-F; Size: 100 ug
Catalog No. 50-167-058