Learn More
Biolegend Purified anti-human CD24
Supplier:  Biolegend 311102
Encompass_Preferred
Purified anti-human CD24 [ML5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee ; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-166-603