Learn More
Biolegend Purified anti-human CD25
Supplier:  Biolegend 302602
Encompass_Preferred
Purified anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-165-875