Learn More
Biolegend Purified anti-human CD253 (Trail)
Supplier:  Biolegend 308202
Encompass_Preferred
Purified anti-human CD253 (Trail) [RIK-2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC, Block; Size: 100 ug
Catalog No. 50-166-466