Learn More
Biolegend Purified anti-human CD255 (TWEAK)
Catalog No. 50166472
Encompass_Preferred
Purified anti-human CD255 (TWEAK) [CARL-1]; Isotype: Mouse IgG3, κ; Reactivity: Human, African Green, Baboon; Apps: FC, IHC; Size: 100 ug
Catalog No. 50-166-472 Supplier Biolegend Supplier No. 308302
Purified anti-human CD255 (TWEAK) [CARL-1]; Isotype: Mouse IgG3, κ; Reactivity: Human, African Green, Baboon; Apps: FC, IHC; Size: 100 ug