Learn More
Biolegend Purified anti-human CD262 (DR5, TRAIL-R2)
Supplier:  Biolegend 307402
Encompass_Preferred
Purified anti-human CD262 (DR5, TRAIL-R2) [DJR2-4 (7-8)]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-166-407