Learn More
Biolegend Purified anti-human CD270 (HVEM, TR2)
Supplier:  Biolegend 318802
Encompass_Preferred
Purified anti-human CD270 (HVEM, TR2) [122]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, WB, ELISA; Size: 100 ug
Catalog No. 50-166-988