Learn More
Biolegend Purified anti-human CD28
Supplier:  Biolegend 302902
Encompass_Preferred
Purified anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC, IHC-F, Costim, FA; Size: 100 ug
Catalog No. 50-165-915