Learn More
Biolegend Purified anti-human CD283 (TLR3)
Supplier:  Biolegend 315002
Encompass_Preferred
Purified anti-human CD283 (TLR3) [TLR-104]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC, WB; Size: 100 ug
Catalog No. 50-166-824