Learn More
Biolegend Purified anti-human CD29
Supplier:  Biolegend 303001
Encompass_Preferred
Purified anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC, ICC, IHC, IP; Size: 25 ug
Catalog No. 50-165-937