Learn More
Biolegend Purified anti-human CD3
Supplier:  Biolegend 344801
Encompass_Preferred
Purified anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ICC, IHC-F, WB; Size: 25 ug
Catalog No. 50-168-296