Learn More
Biolegend Purified anti-human CD3
Supplier:  Biolegend 317301
Encompass_Preferred
Purified anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-166-904