Learn More
Biolegend Purified anti-human CD314 (NKG2D)
Supplier:  Biolegend 320802
Encompass_Preferred
Purified anti-human CD314 (NKG2D) [1D11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IP; Size: 100 ug
Catalog No. 50-167-036