Learn More
Biolegend Purified anti-human CD321 (F11R)
Supplier:  Biolegend 353502
Encompass_Preferred
Purified anti-human CD321 (F11R) [OV-5B8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-168-794