Learn More
Biolegend Purified anti-human CD325 (N-Cadherin)
Supplier:  Biolegend 350802
Encompass_Preferred
Purified anti-human CD325 (N-Cadherin) [8C11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC, WB, Block; Size: 100 ug
Catalog No. 50-168-623