Learn More
Biolegend Purified anti-human CD33
Supplier:  Biolegend 303302
Encompass_Preferred
Purified anti-human CD33 [HIM3-4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-605-240