Learn More
Biolegend Purified anti-human CD334 (FGFR4)
Supplier:  Biolegend 324302
Encompass_Preferred
Purified anti-human CD334 (FGFR4) [4FR6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-167-240