Learn More
Biolegend Purified anti-human CD336 (NKp44)
Supplier:  Biolegend 325102
Encompass_Preferred
Purified anti-human CD336 (NKp44) [P44-8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, Block; Size: 100 ug
Catalog No. 50-167-264