Learn More
Biolegend Purified anti-human CD337 (NKp30)
Supplier:  Biolegend 325202
Encompass_Preferred
Purified anti-human CD337 (NKp30) [P30-15]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, ELISA, Block; Size: 100 ug
Catalog No. 50-167-272