Learn More
Biolegend Purified anti-human CD38
Supplier:  Biolegend 303502
Encompass_Preferred
Purified anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC, CyTOF, IHC-F; Size: 100 ug
Catalog No. 50-165-993