Learn More
Biolegend Purified anti-human CD40
Supplier:  Biolegend 334302
Encompass_Preferred
Purified anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey ; Apps: FC, Costim; Size: 100 ug
Catalog No. 50-167-768