Learn More
Biolegend Purified anti-human CD41
Supplier:  Biolegend 303702
Encompass_Preferred
Purified anti-human CD41 [HIP8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Cynomolgus, Rhesus; Apps: FC, CyTOF, IHC-F; Size: 100 ug
Catalog No. 50-166-024