Learn More
Biolegend Purified anti-human CD42b
Supplier:  Biolegend 303902
Encompass_Preferred
Purified anti-human CD42b [HIP1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, Block, IHC-F, WB; Size: 100 ug
Catalog No. 50-166-040