Learn More
Biolegend Purified anti-human CD43
Supplier:  Biolegend 343202
Encompass_Preferred
Purified anti-human CD43 [CD43-10G7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC, IP; Size: 100 ug
Catalog No. 50-168-167