Learn More
Biolegend Purified anti-human CD45
Supplier:  Biolegend 304002
Encompass_Preferred
Purified anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, CyTOF, IHC-P, WB, IHC-F, ICC; Size: 100 ug
Catalog No. 50-166-049